missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD6/PSR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71693
This item is not returnable.
View return policy
Description
JMJD6/PSR Polyclonal specifically detects JMJD6/PSR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| JMJD6/PSR | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide | |
| bifunctional arginine demethylase and lysyl-hydroxylase JMJD6, EC 1.14.11, EC 1.14.11.-, Histone arginine demethylase JMJD6, JmjC domain-containing protein 6, jumonji domain containing 6, Jumonji domain-containing protein 6, KIAA0585PSR, Lysyl-hydroxylase JMJD6, Peptide-lysine 5-dioxygenase JMJD6, Phosphatidylserine receptorPTDSR, Protein PTDSR, PTDSR1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen has 98% identity with mouse, rat and bovine. Immunogen has 91% identity to Xenopus and Zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Q6NYC1 | |
| JMJD6 | |
| A genomic peptide made to an internal region of the human JMJD6 protein (within residues 150-350). [Swiss-Prot Q6NYC1] | |
| 0.05 mL | |
| Epigenetics, Stem Cell Signaling Pathway | |
| 23210 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction