missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JMJD6 Polyclonal Antibody

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148556

Product Code. 30533980

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a nuclear protein with a JmjC domain. JmjC domain-containing proteins are predicted to function as protein hydroxylases or histone demethylases. This protein was first identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells; however, subsequent studies have indicated that it does not directly function in the clearance of apoptotic cells, and questioned whether it is a true phosphatidylserine receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JMJD6
Polyclonal
Unconjugated
JMJD6
5730436I23Rik; apoptotic cell clearance receptor PtdSerR; arginine demethylase and lysine hydroxylase; bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; D11Ertd195e; Histone arginine demethylase JMJD6; jmjC domain-containing protein 6; Jmjd6; jumonji domain containing 6; jumonji domain-containing protein 6; KIAA0585; Lysyl-hydroxylase JMJD6; mKIAA0585; Peptide-lysine 5-dioxygenase JMJD6; phosphatidylserine receptor; Protein PTDSR; PSR; PtdSerR; Ptdsr; PTDSR1
Goat
Antigen affinity chromatography
RUO
107817
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9ERI5
JMJD6
Mouse myeloma cell line NS0-derived recombinant mouse JMJD6/PSR, Ala289-Arg403.
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.