missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | JMJD4 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16891165
|
Proteintech
21572-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16881165
|
Proteintech
21572-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The jumonji domain-containing protein (JMJD) family is defined by the presence of the JmjC domain that is observed in several diverse species. While several JMJD proteins have been identified as being involved in chromatin regulation, histone demethylation and development, the JMJD4 has not been identified. JMJD4 is expressed in multiple tissues and At least three isoforms of JMJD4 are known to exist.Specifications
| JMJD4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H9V9 | |
| 65094 | |
| JMJD4 Fusion Protein Ag16099 | |
| Primary | |
| -20°C | |
| JMJD4 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| JMJD4, jumonji domain containing 4 | |
| JMJD4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title