missing translation for 'onlineSavingsMsg'
Learn More

JMJD4 Antibody (JMJD4/32b/H12) - BSA Free, Novus Biologicals™

Product Code. 18621808 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18621808 0.025 mL 0.03mL
18655817 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18621808 Supplier Novus Biologicals Supplier No. NBP2504250.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

JMJD4 Monoclonal antibody specifically detects JMJD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD4
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone JMJD4/32b/H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias FLJ12517, jumonji domain containing 4, jumonji domain-containing protein 4
Host Species Mouse
Immunogen Recombinant protein
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 65094
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.