missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ JMJD2D Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 55693 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-03357 |
| Gene Alias | EC 1.14.11, EC 1.14.11.-, FLJ10251, JHDM3D, JmjC domain-containing histone demethylation protein 3 D, JMJD2Dlysine-specific demethylase 4 D, jumonji domain containing 2 D, Jumonji domain-containing protein 2 D, lysine (K)-specific demethylase 4 D, MGC141909 |
| Gene Symbol | KDM4D |
| Quantity | 0.1 mg |
| Research Category | Transcription Factors and Regulators |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction