missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115461
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KDM4C / GASC1 / JMJD2C.
Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 'Lys-9' and H3 'Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
Specifications
| JMJD2C | |
| Polyclonal | |
| Unconjugated | |
| KDM4C | |
| 2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; JMJD2C; jumonji domain containing 2C; jumonji domain-containing protein 2C; Kdm4c; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23081, 298144, 76804 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8VCD7, Q9H3R0 | |
| KDM4C | |
| A synthesized peptide derived from human KDM4C(Accession Q9H3R0), corresponding to amino acid residues Y472-G522. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction