missing translation for 'onlineSavingsMsg'
Learn More

JMJD1C Antibody, DyLight 488, Novus Biologicals™

Product Code. 18420075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420075 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420075 Supplier Novus Biologicals Supplier No. NBP177072G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JMJD1C Polyclonal antibody specifically detects JMJD1C in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1C
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp761F0118, EC 1.14.11, EC 1.14.11.-, FLJ14374, JHDM2C, jumonji domain containing 1C, Jumonji domain-containing protein 1C, KIAA1380TR-interacting protein 8, probable JmjC domain-containing histone demethylation protein 2C, RP11-10C13.2, thyroid hormone receptor interactor 8, thyroid receptor interacting protein 8, Thyroid receptor-interacting protein 8, TRIP8, TRIP-8
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide from near the amino terminus of human JMJD1C. The immunogen is located within amino acids 380 - 430 of JMJD1C. Amino Acid Squence: SEQKPENELKNKNTSKINGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221037
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.