missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553459
This item is not returnable.
View return policy
Description
Immunogen sequence: GEVDSNGSDG GEASRGPWKG GNASGEPGLD QRAKQPPSTF VPQINRNIRF ATYTKENGRT LVVQDEPVGG DTPASFTPYS TATGQTPLAP EVGGAENKEA GKTLEQVGQG IVASAAVVTT ASSTPNTVRI SDTGLAAGTV PE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
Specifications
| JMJD1B | |
| Polyclonal | |
| Unconjugated | |
| KDM3B | |
| 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JmjC domain-containing histone demethylation protein 2B; JMJD1B; jumonji domain containing 1B; jumonji domain-containing protein 1B; KDM3B; KDM3B lysine (K)-specific demethylase 3B; KIAA1082; lysine (K)-specific demethylase 3B; lysine demethylase 3B; lysine-specific demethylase 3B; mKIAA1082; NET22; Nuclear protein 5qNCA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 277250, 51780, 682469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZPY7, Q7LBC6 | |
| KDM3B | |
| Recombinant protein corresponding to Human JMJD1B. Recombinant protein control fragment (Product #RP-91620). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction