missing translation for 'onlineSavingsMsg'
Learn More

JMJD1B Antibody, DyLight 680, Novus Biologicals™

Product Code. 18411783 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411783 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411783 Supplier Novus Biologicals Supplier No. NBP177071FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JMJD1B Polyclonal antibody specifically detects JMJD1B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1B
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias C5orf7, EC 1.14.11, EC 1.14.11.-, JHDM2B, JmjC domain-containing histone demethylation protein 2B, JMJD1B, jumonji domain containing 1B, Jumonji domain-containing protein 1B, KIAA1082chromosome 5 open reading frame 7, lysine (K)-specific demethylase 3B, lysine-specific demethylase 3B, NET22,5qNCA, Nuclear protein 5qNCA
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide from near the center of human JMJD1B. The immunogen is located within amino acids 1050 - 1100 of JMJD1B. Amino Acid Squence: YRLRKSRPRSETEEMGDEE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51780
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.