missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104455
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total JM4.
JM4 is a 19 kDa four-transmembrane protein binding to the CCR5 receptor. It plays a role in the regulation of intracellular protein transport. It is expressed in normal brain, small intestine, lung, spleen, and pancreas and has also been detected in tumors of breast, colon, lung, and ovary.
Specifications
| JM4 | |
| Polyclonal | |
| Unconjugated | |
| PRAF2 | |
| AA986329; C78013; DXImx39e; Jena-Muenchen 4; JM4; PRA1 domain family 2; PRA1 domain family member 2; PRA1 domain family, member 2; PRA1 family protein 2; Praf2; RGD1566287; Sfc20; Yip6a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11230, 367743, 54637 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60831, Q9JIG8 | |
| PRAF2 | |
| A synthesized peptide derived from human PRAF2(Accession O60831), corresponding to amino acid residues K146-S178. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction