missing translation for 'onlineSavingsMsg'
Learn More

JM-21 Multi-element Oil Based Standard, Specpure™ 900μg/g, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Alfa Aesar 036769.22

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15496165

  • £643.00 / 100g
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

JM-21 Multi-element Oil Based Standard
Matrix: Hydrocarbon oil
MFCD00151262
Liquid
900μg/g
100 g
Specpure™
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only