missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JLP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111525
This item is not returnable.
View return policy
Description
Immunogen sequence: NLSGGKTRDG GSVVGASVFY KDVAGLDTEG SKQRSASQSS LDKLDQELKE QQKELKNQEE LSSLVWICTS THSATKVL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 91%.
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
Specifications
| JLP | |
| Polyclonal | |
| Unconjugated | |
| SPAG9 | |
| 3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; Cancer/testis antigen 89; c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; CT89; HLC4; HLC6; HLC-6; HSS; Human lung cancer oncogene 6 protein; Jip4; JIP-4; JLP; JNK interacting protein; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; Jsap2; JSAP2a; Kiaa0516; lung cancer oncogene 4; Mapk8ip4; Max-binding protein; mitogen-activated protein kinase 8-interacting protein 4; PHET; PIG6; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; SPAG9; sperm associated antigen 9; sperm associated antigen 9 variant 5; sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1; syd1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60271 | |
| SPAG9 | |
| Recombinant Protein corresonding to Human JLP. Recombinant protein control fragment (Product #RP-107069). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction