missing translation for 'onlineSavingsMsg'
Learn More

JK184, MedChemExpress

Product Code. 16496156
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496156 100 mg 100mg
16488327 5 mg 5mg
16435638 10 mg 10mg
16411807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496156 Supplier Medchem Express Supplier No. HY13307
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress JK184 is a potent Hedgehog (Hh) pathway inhibitor with IC50 of 30 nM in mammalian cells.

JK184 is a potent Hedgehog (Hh) pathway inhibitor with IC50 of 30 nM in mammalian cells.

Specifications

Chemical Name or Material JK184
CAS 315703-52-7
Color Pink
Purity Grade Notes Research
Molecular Formula C19H18N4OS
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (142.68 mM)
SMILES CC1=C(C2=CSC(NC3=CC=C(OCC)C=C3)=N2)N4C=CC=CC4=N1
Molecular Weight (g/mol) 350.44
Formula Weight 350.44
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.