missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JIP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564408
This item is not returnable.
View return policy
Description
Immunogen sequence: AGVNLSGWRP NEDDAGNGVK PAPGRDPLTC DREGDGEPKS AHTSPEKKKA KELPEMDATS SRVWILTSTL TTSKVVIIDA NQPGTVVDQF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
c-Jun NH2-terminal kinases (JNKs) are distant members of the MAP kinase family. JNK1 is activated by dual phosphorylation at a Thr-Pro-Tyr motif in response to ultraviolet (UV) light, and it functions to phosphorylate c-Jun at amino terminal serine regulatory sites Ser 63 and Ser 73, resulting in transcriptional activation. Two additional JNK family members, JNK2 and JNK3, have been identified. JIP-1 (for JNK interacting protein-1) has been identified as a cytoplasmic inhibitor of JNK that retains JNK in the cytoplasm, thereby inhibiting JNK-regulated gene expression. Evidence suggests that JNK1 and JNK2 bind to JIP-1 with greater affinity than to ATF-2 and c-Jun, which are targets of the JNK signaling pathway. JIP-1 contains an amino terminal JNK binding domain and a carboxy terminal SH3 domain. ATF-2 and c-Jun also contain the JNK binding domain and are thought to compete with JIP-1 for JNK binding. Multiple splice variants of JIP-1, including JIP-1b, JIP-1c (also designated islet-brain 1 or IB-1), JIP-2a, JIP-2b and JIP-3, have been identified in brain.
Specifications
| JIP3 | |
| Polyclonal | |
| Unconjugated | |
| MAPK8IP3 | |
| BB120594; c-Jun NH2-terminal kinase (JNK)/stress-activated protein kinase-associated protein 1; C-jun-amino-terminal kinase interacting protein 3; C-Jun-amino-terminal kinase-interacting protein 3; D17Wsu15e; homolog of Drosophila Sunday driver 2; JIP3; JIP-3; JNK MAP kinase scaffold protein 3; JNK/SAPK-associated protein 1; JNK/SAPK-associated protein 1c; JNK/SAPK-associated protein-1; JNK/stress-activated protein kinase-associated protein 1; JNK-interacting protein 3; JSAP1; JSAP1a; JSAP1b; JSAP1c; JSAP1d; JUN/SAPK-associated protein 1; KIAA1066; MAPK8IP3; mitogen activated protein kinase 8 interacting protein 3; mitogen-activated protein kinase 8 interacting protein 3; mitogen-activated protein kinase 8-interacting protein 3; mKIAA1066; Sunday driver 2; syd; SYD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPT6 | |
| MAPK8IP3 | |
| Recombinant protein corresponding to Human JIP3. Recombinant protein control fragment (Product #RP-104067). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction