missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
JIP1 Monoclonal antibody specifically detects JIP1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | JIP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8H5 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C-Jun-amino-terminal kinase-interacting protein 1;IB1;JIP1;JIP-1;MAPK8IP1;PRKM8IP |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human MAPK8IP1 (NP_005447) produced in HEK293T cell. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?