missing translation for 'onlineSavingsMsg'
Learn More

JIP1 Antibody (OTI13F10) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30687540 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687540 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687540 Supplier Novus Biologicals Supplier No. NBP345039AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

JIP1 Monoclonal antibody specifically detects JIP1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen JIP1
Applications Western Blot
Classification Monoclonal
Clone OTI13F10
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias C-Jun-amino-terminal kinase-interacting protein 1;IB1;JIP1;JIP-1;MAPK8IP1;PRKM8IP
Host Species Mouse
Immunogen Full length human recombinant protein of human MAPK8IP1 (NP_005447) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9479
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.