missing translation for 'onlineSavingsMsg'
Learn More

Jionoside B1, MedChemExpress

Product Code. 16481688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481688 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481688 Supplier Medchem Express Supplier No. HYN2218
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Jionoside B1 is a phenylpropanoid isolated from herbs of Eriophyton wallichii.

Jionoside B1 is a phenylpropanoid isolated from herbs of Eriophyton wallichii.

Specifications

Chemical Name or Material Jionoside B1
CAS 120406-37-3
Color White
Purity Grade Notes Research
Molecular Formula C37H50O20
Quantity 5 mg
SMILES O=C(/C=C/C1=CC(OC)=C(O)C=C1)O[C@H]([C@H](O[C@H]2OCCC3=CC(O)=C(OC)C=C3)CO[C@@H]([C@@H]([C@@H](O)[C@H]4O)O)O[C@@H]4CO)[C@@H]([C@H]2O)O[C@@](O[C@@H](C)[C@H](O)[C@H]5O)([H])[C@@H]5O
Molecular Weight (g/mol) 814.78
Formula Weight 814.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.