missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JIK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582574
This item is not returnable.
View return policy
Description
Immunogen sequence: ESQEDEEDSE HGTSLNREMD SLGSNHSIPS MSVSTGSQSS SVNSMQEVMD ESSSELVMMH DDESTINSSS SVVHKKDHVF IRDEAGHGDP RPEPRPTQSV QSQALHYRNR Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 85%.
TAOK3 (JIK) is a serine/threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family. JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.
Specifications
| JIK | |
| Polyclonal | |
| Unconjugated | |
| TAOK3 | |
| 2900006A08Rik; A130052D22; A430105I05Rik; Arg357; Axotomy-related gene 357 protein; CTCL-associated antigen HD-CL-09; Cutaneous T-cell lymphoma-associated antigen HD-CL-09; dendritic cell-derived protein kinase; DKFZp666H245; DPK; EC 2.7.11.1; FLJ31808; hKFC-A; JIK; JNK/SAPK-inhibitory kinase; Jun kinase-inhibitory kinase; KDS; Kinase from chicken homolog A; MAP3K18; Serine/threonine-protein kinase TAO3; STE20-like kinase; STE2-like kinase; tao; TAO kinase 3; Taok3; Thousand and one amino acid protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2K8 | |
| TAOK3 | |
| Recombinant protein corresponding to Human TAOK3. Recombinant protein control fragment (Product #RP-89823). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction