missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JIK Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97320AF750
This item is not returnable.
View return policy
Description
JIK Polyclonal antibody specifically detects JIK in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| JIK | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| CTCL-associated antigen HD-CL-09, Cutaneous T-cell lymphoma-associated antigen HD-CL-09, Dendritic cell-derived protein kinase, DPKJNK/SAPK-inhibitory kinase, EC 2.7.11, FLJ31808, hKFC-A, JIKEC 2.7.11.1, Jun kinase-inhibitory kinase, KDS, Kinase from chicken homolog A, MAP3K18DKFZp666H245, serine/threonine-protein kinase TAO3, STE20-like kinase, TAO kinase 3, Thousand and one amino acid protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human JIK fragment. | |
| 0.1 mL | |
| Cell Biology, Protein Kinase, Signal Transduction | |
| 51347 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction