missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Jiffy airkraft bags, 245mm x 170mm
Brand: Manutan A329212
This item is not returnable.
View return policy
Description
Ideal for high volume mailings with secure peel and seal closure. Protective air filled bubble lining keeping contents safe. 92gsm, tear and puncture-resistant.- Bag no. : JL1
- Internal length : 245mm
- Internal width : 170mm