missing translation for 'onlineSavingsMsg'
Learn More

JI-101, MedChemExpress

Product Code. 16489557
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489557 5 mg 5mg
16447076 100 mg 100mg
16443067 50 mg 50mg
16437018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489557 Supplier Medchem Express Supplier No. HY16265
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress JI-101 is an orally available multi-kinase inhibitor of VEGFR2, PDGFRβ and EphB4 with potent anti-cancer activity.

JI-101 is an orally available multi-kinase inhibitor of VEGFR2, PDGFRβ and EphB4 with potent anti-cancer activity.

Specifications

Chemical Name or Material JI-101
CAS 900573-88-8
Color White
Purity Grade Notes Research
Molecular Formula C22H20BrN5O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (214.44 mM)
SMILES O=C(NC1=CC(Br)=CC=C1OC)NC2=CC=CC3=C2C=CN3CC4=CC(N)=NC=C4
Molecular Weight (g/mol) 466.33
Formula Weight 466.33
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.