missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JHU 37152
GREENER_CHOICE

Product Code. 18749286
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749286 10 mg 10mg
18708886 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18749286 Supplier Tocris Bioscience™ Supplier No. 7197/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and highly potent hM3Dq and hM4Di DREADD agonist; blood brain barrier penetrant

JHU 37152 is a high affinity and highly potent activator of hM3Dq and hM4Di DREADDs (Ki values are 1.8 nM and 8.7 nM for hM3Dq and hM4Di in vitro, respectively; EC50 values are 5 nM and 0.5 nM for hM3Dq and hM4Di in vitro, respectively). Selectively displaces [3H]clozapine from DREADDs in vivo, but not from other clozapine binding sites. Inhibits locomotor activity in mice expressing hM3Dq and hM4Di in D1-expressing neurons. Brain penetrant in mice, rats and non-human primates.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-11-(4-ethylpiperazin-1-yl)-1-fluoro-5 H-dibenzo[b,e][1,4 ]diazepine
CAS 2369979-67-7
Quantity 10 mg
Target DREADD Activators
Molecular Formula C19H20ClFN4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.