missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JH-XI-10-02
GREENER_CHOICE

Product Code. 18755606
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18755606 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18755606 Supplier Tocris Bioscience™ Supplier No. 7304/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CDK8 Degrader (PROTAC™)

JH-XI-10-02 is a CDK8 Degrader comprising the cereblon binding ligand Pomalidomide (Cat. No. 6302) joined by a linker to a CDK8 inhibitor. JH-XI-10-02 (1 μM) induces significant proteasome-dependent degradation of CDK8 in Jurkat cells after 24 h, with no significant effect on CDK19 levels.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 15-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]amino]-N-[(3β,5α,17β)-17-(7-isoquinolinyl)androstan-3-yl]-N-methyl-4,7,10,13-tetraoxapentadecanamide
CAS 2209085-22-1
Quantity 2 mg
Target Active Degraders
Molecular Formula C53H69N5O9
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.