missing translation for 'onlineSavingsMsg'
Learn More

JH-II-127, TRC

Product Code. 30391227
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391227 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391227 Supplier Toronto Research Chemicals Supplier No. TRCJ21130310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material JH-II-127
CAS 1700693-08-8
Molecular Formula C19H21ClN6O3
Quantity 10 mg
Synonym [4-[[5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-4-morpholinyl-methanone
InChI Formula InChI=1S/C19H21ClN6O3/c1-21-16-15-12(20)10-22-17(15)25-19(24-16)23-13-4-3-11(9-14(13)28-2)18(27)26-5-7-29-8-6-26/h3-4,9-10H,5-8H2,1-2H3,(H3,21,22,23,24,25)
SMILES O=C(C1=CC(OC)=C(C=C1)NC2=NC(NC)=C3C(Cl)=CNC3=N2)N4CCOCC4
IUPAC Name [4-[[5-chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-morpholin-4-ylmethanone
Molecular Weight (g/mol) 416.87
Formula Weight 416.1364
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.