missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JH 295

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
JH 295 is an irreversible Nek2 inhibitor (IC50=770 nM). Inhibits Nek2 via alkylation of residue Cys22. Displays 5-fold selectivity for Nek2 over Cdk1. Exhibits no significant effect on Aurora B or Plk1 kinases, bipolar spindle assembly or spindle assembly checkpoint. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | N-[(3Z)-3-[(2-Ethyl-4-methyl-1H-imidazol-5-yl)methylene]-2,3-dihydro-2-oxo-1H-indol-5-yl]-2-propynamide |
| CAS | 1311143-71-1 |
| Assay Percent Range | >98% |
| Quantity | 2 mg |
| Molecular Formula | C18H16N4O2 |
| Formula Weight | Observed MW: 320.35 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction