missing translation for 'onlineSavingsMsg'
Learn More

JG-98, MedChemExpress

Product Code. 16451127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451127 50 mg 50mg
16417627 5 mg 5mg
16404878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451127 Supplier Medchem Express Supplier No. HY117282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages.

JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages.

Specifications

Chemical Name or Material JG-98
Physical Form Solid
CAS 1456551-16-8
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H21Cl2N3OS3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 5 mg/mL (9.35 mM; Need ultrasonic)
SMILES CN1/C(SC2=CC(Cl)=CC=C12)=C3S/C(N(CC)C\3=O)=C\C4=[N+](C=CS4)CC5=CC=CC=C5.[Cl-]
Molecular Weight (g/mol) 534.54
Formula Weight 534.54
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.