missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ JF-NP-26
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
JF-NP-26 is a caged Raseglurant (Cat. No. 4416). Releases raseglurant upon exposure to 405 nm (violet) light. Inhibits the effects of Quisqualate (Cat. No. 0188) in cultured neurons when exposed to 405 nm light. Exhibits analgesic activity in pain models in vivo on photoactivation. Quantum yield = 0.18.
Specifications
Specifications
| Chemical Name or Material | (7-(Diethylamino)-2-oxo-2H-chromen-4-yl)methyl (2-((3-fluorophenyl)ethynyl)-4,6-dimethylpyridin-3-yl)carbamate |
| CAS | 2341841-03-8 |
| Quantity | 5 mg |
| Molecular Formula | C30H28FN3O4 |
| Formula Weight | 513.56 |
| Physical Form | Solid |
| Color | Pale-yellow |
| Product Type | Caged Raseglurant |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction