Learn More
Fisherbrand™ Jewellery Clamp
Click to view available options
For Use With (Equipment):
Steddy Microscopes
Unit Size:
Each
Description
- Jewellery Clamps
- For STEDDY-T Trinocular Stereo Microscope FB69056
The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
Robust stereo zoom microscope
- Integral single piece column stand eliminates vibration and image distortion
- Ergonomic diamond shaped base
- Total magnification range 2.5x to 120x
- Variable intensity transmitted and incident light sources
Specifications
Specifications
| Type | Jewelry Clamp |
| For Use With (Equipment) | Steddy Microscopes |
| Quantity | 1 Ea. |