Learn More
Invitrogen™ JetQuick™ Blood and Cell Culture DNA Midiprep Kit
Beschreibung
Utilizes silica membrane-based technology that allows for isolation of 20-50 kb DNA that is suitable for a wide range of downstream applications, including PCR, restriction enzyme digestion, and Southern blotting.
Key benefits include:
• Large volume processing, up to 3 mL of blood or saliva
• Long double-stranded DNA (20-50 kb) recovery
• Compatible with centrifugation or a vacuum manifold
The midiprep format enables the processing of up to 3 mL of whole blood or other body fluid. DNA from up to 1 x 108 mammalian cells can be isolated. The purified DNA is ready for downstream analysis.
Spezifikation
Spezifikation
| Content And Storage | • 1 x 165 mL Buffer K1 x Store at Room Temperature • 2 x 87 mL Buffer K2 x Store at Room Temperature • 2 x 124 mL Buffer KX, Store at Room Temperature • 1 x 110 mL Elution Buffer, Store at Room Temperature • 4 x 1.3 mL Rnase A, Store 2°C to 8°C • 1 x 320 mg Protease (lyophilized powder), Store 2°C to 8°C • 10 Spin Column Bags (5 columns/bag) |
| Format | Kit |
| Isolation Technology | Spin Column |
| Scale | Midi |
| Sample Type | Blood, Cells |
| For Use With (Application) | Real-Time PCR, PCR |
| High-throughput Compatibility | Not High-throughput Compatible |
| No. of Reactions | 50 Preps |
| Product Line | JetQuick |
| Product Type | Blood and Cell Culture DNA Kit |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.