Learn More
Invitrogen™ JetQuick™ Blood and Cell Culture DNA Maxiprep Kit
Description
Utilizes silica membrane-based technology that allows for isolation of 20-50 kb DNA that is suitable for a wide range of downstream applications, including PCR, restriction enzyme digestion, and Southern blotting.
Key benefits include:
• Large volume processing, up to 10 mL of blood or saliva
• Long double-stranded DNA (20-50 kb) recovery
• Compatible with centrifugation or a vacuum manifold
The maxiprep format enables the processing of up to 10 mL of whole blood or other body fluid. DNA from up to 5 x 108 mammalian cells can be isolated. The purified DNA is ready for downstream analysis.
Specifications
Specifications
| Content And Storage | • 3 x 185 mL Buffer K1 x Store at Room Temperature • 2 x 87 mL Buffer K2 x Store at Room Temperature • 2 x 124 mL Buffer KX, Store at Room Temperature • 1 x 110 mL Elution Buffer, Store at Room Temperature • 13 x 1.3 mL Rnase A, Store 2°C to 8°C • 1 x 560 mg Protease (lyophilized powder), Store 2°C to 8°C • 10 Spin Column Bags (5 columns/bag) |
| Format | Kit |
| Isolation Technology | Spin Column |
| Scale | Maxi |
| Sample Type | Blood, Cells |
| For Use With (Application) | Real-Time PCR, PCR |
| High-throughput Compatibility | Not High-throughput Compatible |
| No. of Reactions | 50 Preps |
| Product Line | JetQuick |
| Product Type | Blood and Cell Culture DNA Kit |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.