Learn More
Invitrogen™ JetQuick™ Blood and Cell Culture DNA Maxiprep Kit
Description
Utilizes silica membrane-based technology that allows for isolation of 20-50 kb DNA that is suitable for a wide range of downstream applications, including PCR, restriction enzyme digestion, and Southern blotting.
Key benefits include:
• Large volume processing, up to 10 mL of blood or saliva
• Long double-stranded DNA (20-50 kb) recovery
• Compatible with centrifugation or a vacuum manifold
The maxiprep format enables the processing of up to 10 mL of whole blood or other body fluid. DNA from up to 5 x 108 mammalian cells can be isolated. The purified DNA is ready for downstream analysis.
Specifications
Specifications
| Content And Storage | • 1 x 220 mL Buffer K1 x Store at Room Temperature • 1 x 72 mL Buffer K2 x Store at Room Temperature • 1 x 105 mL Buffer KX, Store at Room Temperature • 1 x 44 mL Elution Buffer, Store at Room Temperature • 5 x 1.3 mL Rnase A, Store 2°C to 8°C • 5 x 220 mg Protease (lyophilized powder), Store 2°C to 8°C • 4 x Spin Column Bags (5 columns/bag) |
| Format | Kit |
| Isolation Technology | Spin Column |
| Scale | Maxi |
| Sample Type | Blood, Cells |
| For Use With (Application) | Real-Time PCR, PCR |
| High-throughput Compatibility | Not High-throughput Compatible |
| No. of Reactions | 20 Preps |
| Product Line | JetQuick |
| Product Type | Blood and Cell Culture DNA Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.