missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
JDP2 Polyclonal specifically detects JDP2 in Human, Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | JDP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | Jun dimerization protein 2, JUNDM2, progesterone receptor co-activator |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-174 of human JDP2 (NP_001128521.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?