missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JDP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563740
This item is not returnable.
View return policy
Description
Immunogen sequence: HRPTCIVRTD SVKTPESEGN PLLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Component of the AP-1 transcription factor that represses transactivation mediated by the Jun family of proteins. Involved in a variety of transcriptional responses associated with AP-1 such as UV-induced apoptosis, cell differentiation, tumorigenesis and antitumogeneris. Can also function as a repressor by recruiting histone deacetylase 3/HDAC3 to the promoter region of JUN. May control transcription via direct regulation of the modification of histones and the assembly of chromatin.
Specifications
| JDP2 | |
| Polyclonal | |
| Unconjugated | |
| JDP2 | |
| c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122953 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WYK2 | |
| JDP2 | |
| Recombinant protein corresponding to Human JDP2. Recombinant protein control fragment (Product #RP-101814). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction