missing translation for 'onlineSavingsMsg'
Learn More

JD-5037, MedChemExpress

Product Code. 16477588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477588 10 mg 10mg
16473607 50 mg 50mg
16457516 100 mg 100mg
16430068 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477588 Supplier Medchem Express Supplier No. HY18697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JD-5037 is a potent CB1R antagonist with an IC50 of 1.5 nM.

JD-5037 is a potent CB1R antagonist with an IC50 of 1.5 nM.

Specifications

Chemical Name or Material JD-5037
CAS 1392116-14-1
Color White
Purity Grade Notes Research
Molecular Formula C27H27Cl2N5O3S
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (436.67 mM; Need ultrasonic)
SMILES NC([C@H](C(C)C)/N=C(NS(=O)(C1=CC=C(Cl)C=C1)=O)\N2N=C(C3=CC=C(Cl)C=C3)[C@@H](C4=CC=CC=C4)C2)=O
Molecular Weight (g/mol) 572.51
Formula Weight 572.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.