Learn More
PSF Progressive Safety™ JCB 4x4 Honey Boot
Product Code. 12862567
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
13
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12862567 6 1 pair
12872567 7 -
12882567 8 1 pair
12892567 9 1 pair
12822567 10 1 pair
12832567 11 -
12842567 12 -
12852567 13 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12862567 Supplier PSF Progressive Safety™ Supplier No. 4X4H6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honey nubuck boot
  • Water resistant upper
  • Waterproof and breathable lining
  • Padded tongue and collar
  • Antistatic
  • Steel toecap and midsole
  • Wider fit
  • TPU chassis with anti-twist technology
  • Lightweight dual density EVA and rubber sole
  • SRA slip resistant
Specifications
Size 6