missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAZF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541686
This item is not returnable.
View return policy
Description
Peptide sequence: KKRYKNVNGI KYHAKNGHRT QIRVRKPFKC RCGKSYKTAQ GLRHHTINFH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
JAZF1 is a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors.This gene encodes a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| JAZF1 | |
| Polyclonal | |
| Unconjugated | |
| JAZF1 | |
| AI591476; C820002C15; DKFZp761K2222; Jaz1; JAZF zinc finger 1; Jazf1; juxtaposed with another zinc finger gene 1; juxtaposed with another zinc finger protein 1; TAK1-interacting protein 27; TIP27; Zinc finger protein 802; ZNF802 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 221895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86VZ6 | |
| JAZF1 | |
| Synthetic peptide directed towards the C-terminal of human JAZF1 (aa 181-230). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction