missing translation for 'onlineSavingsMsg'
Learn More

Jatrorrhizine Chloride, TRC

Product Code. 30400675
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400675 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400675 Supplier Toronto Research Chemicals Supplier No. TRCJ21091010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Jatrorrhizine Chloride
CAS 6681-15-8
Molecular Formula C20 H20 N O4 . Cl
Quantity 10 mg
Synonym Dibenzo[a,g]quinolizinium, 5,6-dihydro-3-hydroxy-2,9,10-trimethoxy-, chloride (1:1), Berbinium, 7,8,13,13a-tetradehydro-3-hydroxy-2,9,10-trimethoxy-, chloride (8CI), Dibenzo[a,g]quinolizinium, 5,6-dihydro-3-hydroxy-2,9,10-trimethoxy-, chloride (9CI), Jatrorrhizine chloride (7CI), NSC 645313, Neprotine chloride
InChI Formula InChI=1S/C20H19NO4.ClH/c1-23-18-5-4-12-8-16-14-10-19(24-2)17(22)9-13(14)6-7-21(16)11-15(12)20(18)25-3;/h4-5,8-11H,6-7H2,1-3H3;1H
SMILES [Cl-].COc1cc2c(CC[n+]3cc4c(OC)c(OC)ccc4cc23)cc1O
IUPAC Name 2,9,10-trimethoxy-5,6-dihydroisoquinolino[3,2-a]isoquinolin-7-ium-3-ol;chloride
Molecular Weight (g/mol) 373.83
Formula Weight 373.1081
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.