missing translation for 'onlineSavingsMsg'
Learn More

Jasmolin I, TRC

Product Code. 30391828
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391828 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391828 Supplier Toronto Research Chemicals Supplier No. TRCJ21050010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Jasmolin I
CAS 4466-14-2
Molecular Formula C21 H30 O3
Quantity 10 mg
Synonym (S)-2-Methyl-4-oxo-3-((Z)-pent-2-en-1-yl)cyclopent-2-en-1-yl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
InChI Formula InChI=1S/C21H30O3/c1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h8-9,11,16,18-19H,7,10,12H2,1-6H3/b9-8-/t16-,18+,19+/m1/s1
SMILES CC\C=C/CC1=C(C)[C@H](CC1=O)OC(=O)[C@@H]2[C@@H](C=C(C)C)C2(C)C
IUPAC Name [(1S)-2-methyl-4-oxo-3-[(Z)-pent-2-enyl]cyclopent-2-en-1-yl] (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Molecular Weight (g/mol) 330.46
Formula Weight 330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.