missing translation for 'onlineSavingsMsg'
Learn More

Jasmolin I + Cinerin I (Mixture), TRC

Product Code. 30391071
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391071 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391071 Supplier Toronto Research Chemicals Supplier No. TRCJ21049510MG
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinerin I∣Jasmolin I
CAS 25402-06-6∣4466-14-2
Molecular Formula C20 H28 O3∣C21 H30 O3
Quantity 10 mg
Synonym Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (1S)-3-(2Z)-2-butenyl-2-methyl-4-oxo-2-cyclopenten-1-yl ester, (1R,3R)-, Cinerin I (6CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, 3-(2-butenyl)-2-methyl-4-oxo-2-cyclopenten-1-yl ester, [1R-[1α[S*(Z)],3β]]-, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, ester with 2-(2-butenyl)-4-hydroxy-3-methyl-2-cyclopenten-1-one (8CI), Cinerin, Cinerin I, (S)-2-Methyl-4-oxo-3-((Z)-pent-2-en-1-yl)cyclopent-2-en-1-yl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
InChI Formula InChI=1S/C20H28O3/c1-7-8-9-14-13(4)17(11-16(14)21)23-19(22)18-15(10-12(2)3)20(18,5)6/h7-8,10,15,17-18H,9,11H2,1-6H3/b8-7-/t15-,17+,18+/m1/s1∣InChI=1S/C21H30O3/c1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h8-9,11,16,18-19H,7,10,12H2,1-6H3/b9-8-/t16-,18+,19+/m1/s1
SMILES C\C=C/CC1=C(C)[C@H](CC1=O)OC(=O)[C@@H]2[C@@H](C=C(C)C)C2(C)C∣CC\C=C/CC1=C(C)[C@H](CC1=O)OC(=O)[C@@H]2[C@@H](C=C(C)C)C2(C)C
IUPAC Name Cinerin I∣Jasmolin I
Molecular Weight (g/mol) 316.43∣330.46
Formula Weight 316.2038∣330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.