missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JARID1A, Mouse anti-Human, Clone: 1H2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005927-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: KVEPEVLSTDTQTSPEPGTRMNILPKRTRRVKTQSESGDVSRNTELKKLQIFGAGPKVVGLAMGTKDKEDEVTRRRKVTNRSDAFNMQMRQRKGTLSVNFSpecifications
| JARID1A | |
| Monoclonal | |
| Unconjugated | |
| jumonji, AT rich interactive domain 1A | |
| KDM5A/RBBP2/RBP2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1H2 | |
| In 1x PBS, pH 7.4 | |
| NM_005056 | |
| JARID1A | |
| JARID1A (NP_005047, 191 a.a. ∼ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5927 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction