missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ JAR Nuclear Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
Specifications
Specifications
| Preparation Method | Use it directly for immuno-precipitation, or heat lysate with SDS gel loading buffer to 95C for 5 minutes followed by rapid cooling for western blot application. If dissociating conditions are required, add reducing agent prior to heating. |
| Quantity | 0.05 mg |
| Content And Storage | Store at -80C. Avoid freeze-thaw cycles. |
| Target | JAR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction