missing translation for 'onlineSavingsMsg'
Learn More

JANEX-1, MedChemExpress

Product Code. 16412677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412677 50 mg 50mg
16409157 5 mg 5mg
16406588 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16412677 Supplier Medchem Express Supplier No. HY15508
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2.

JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2.

Specifications

Chemical Name or Material JANEX-1
CAS 202475-60-3
Color Gray
Purity Grade Notes Research
Molecular Formula C16H15N3O3
Quantity 50 mg
Synonym WHI-P131 Jak3 inhibitor I
Solubility Information DMSO : ≥ 100 mg/mL (336.35 mM)
SMILES OC1=CC=C(NC2=C3C=C(OC)C(OC)=CC3=NC=N2)C=C1
Molecular Weight (g/mol) 297.31
Formula Weight 297.31
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.