Learn More
Tocris Bioscience™ Janelia Fluor™ 646b, NHS ester

Description
Key Information: Janelia Fluor™ 646b, NHS ester is a spontaneously blinking fluorescent dye, supplied with an NHS ester reactive group for the labeling of primary amines.
Used for: suitable for DNA labeling using amine-terminated oligonucleotides in RNA-FISH experiments.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: derived from Janelia Fluor™ 646, SE (Cat. No. 6148). Excitation and emission maxima (λ) are 662 nM and 678nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxopyrrolidin-1-yl 3,7-di(azetidin-1-yl)-5,5-dimethyl-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxylate |
| Quantity | 200 μg |
| Target | Amine-Reactive Janelia Fluor Dyes |
| Molecular Formula | C33H33N3O5Si |
| Purity | 0.9 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.