missing translation for 'onlineSavingsMsg'
Learn More

Janelia Fluor 585, SE

Product Code. 18794569
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18794569 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18794569 Supplier Tocris Bioscience Supplier No. 6418/2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Orange dye (Reactive group: NHS ester). Applications: super resolution microscopy, confocal, two-photon excitation and live cell imaging

Orange fluorogenic fluorescent dye; supplied as an NHS ester for coupling to primary amine groups. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Suitable for confocal fluorescent imaging and super resolution microscopy (SRM) techniques, such as dSTORM (live and fixed cells). Cell permeable. Excitation maximum = 585 nm; emission maximum = 609 nm; Quantum yield = 0.78; Max extinction coefficient = 156,000 M-1cm-1 (measured in Ethanol or TFE plus 0.1% TFA); Correction factor = 0.0990. Exhibits low non-specific background staining.

To measure the absorbance spectrum of this dye we recommend the following solvent: trifluoroethanol plus 0.1% TFA.

We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.

HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[10-[2-Carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]-7-(3,3-difluoro-1-azetidinyl)-9,9-dimethyl-2(9 H)-anthracenylidene]-3,3-difluoroazetidinium, inner salt
CAS 1811539-88-4
Quantity 2 mg
Target Amine-Reactive Janelia Fluor™ Dyes
Molecular Formula C34H27F4N3O6
Formula Weight Observed MW: 649.6
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.