missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAM2 Monoclonal Antibody (7E4C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548773
This item is not returnable.
View return policy
Description
JAM2 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs.
Specifications
| JAM2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P57087 | |
| JAM2 | |
| Purified recombinant fragment of human CD322 (AA: extra 29-238) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 7E4C5 | |
| Unconjugated | |
| JAM2 | |
| 1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM | |
| Mouse | |
| Protein G | |
| RUO | |
| 58494 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction