missing translation for 'onlineSavingsMsg'
Learn More

JAM-C Antibody (CRAM-18F26), Novus Biologicals™

Product Code. 18364182 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364182 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364182 Supplier Novus Biologicals Supplier No. NBP311221

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

JAM-C Monoclonal specifically detects JAM-C in Human, Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAM-C
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone CRAM-18F26
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias JAM-2, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C
Host Species Rat
Immunogen The monoclonal antibody CRAM-18 F26 recognizes junctional adhesion molecule-C (JAM-C) also known as JAM-2, a 45 kD cell adhesion molecule (CAM).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83700
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.