missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-B/VE-JAM Antibody (988934) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10743M
This item is not returnable.
View return policy
Description
JAM-B/VE-JAM Monoclonal antibody specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| JAM-B/VE-JAM | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58494 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 988934 | |
| Flow Cytometry, CyTOF-ready | |
| C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322 | |
| Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM, Phe29-Asn236, Accession # P57087 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction