missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-A Rat anti-Mouse, Clone: RM0116-1M21, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22494
This item is not returnable.
View return policy
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot.
Specifications
| JAM-A | |
| Monoclonal | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| 50848 | |
| Mouse | |
| Lyophilized |
| Western Blot | |
| RM0116-1M21 | |
| Western Blot 1:500-1:1000 | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| Mouse recombinant protein of the extracellular domain of JAM-A | |
| 0.1 mg | |
| Primary | |
| Reconstitute with 0.2 mL sterilized PBS to a final concentration of 0.5 mg/mL. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction