missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAKMIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05109-100UL
This item is not returnable.
View return policy
Description
JAKMIP2 Polyclonal specifically detects JAKMIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| JAKMIP2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9832 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| CTCL tumor antigen HD-CL-04, JAMIP2KIAA0555Jak and microtubule interacting protein 2, janus kinase and microtubule interacting protein 2, janus kinase and microtubule-interacting protein 2, NECC1, neuroendocrine long coiled-coil 1, Neuroendocrine long coiled-coil protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 561-810 of human JAKMIP2 (NP_055605.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction