missing translation for 'onlineSavingsMsg'
Learn More

JAKMIP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18432696 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432696 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432696 Supplier Novus Biologicals Supplier No. NBP281959MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JAKMIP1 Polyclonal antibody specifically detects JAKMIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAKMIP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FLJ31564, GABA-B receptor-binding protein, GABABRBP, JAMIP1Jak and microtubule interacting protein 1, janus kinase and microtubule interacting protein 1, marlin-1, MARLIN1janus kinase and microtubule-interacting protein 1, Multiple alpha-helices and RNA-linker protein 1, multiple coiled-coil GABABR1-binding protein
Host Species Rabbit
Immunogen Rabbit polyclonal JAKMIP1 antibody was raised against a 16 amino acid peptide near the center of human JAKMIP1. The immunogen is located within amino acids 260 - 310 of JAKMIP1. Amino Acid Squence: ELMGVQDQHMDERDVR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 152789
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.